Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199593.1 | NP_001186522.1 | 4 | 613 | ||
NM_177262.4 | NP_796236.2 | 4 | 724 | |||
GenBank mRNA | AK089431.1 | - | 4 | 588 | ||
AK134792.1 | - | 5 | 708 | |||
AK149649.1 | - | 4 | 587 | |||
AK150062.1 | - | 4 | 550 | |||
AK170556.1 | - | 4 | 681 | |||
AK171297.1 | - | 4 | 596 | |||
BC052923.1 | - | 4 | 671 | 6312892 |