Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177372.3 | NP_796346.2 | 4 | 608 | ||
GenBank mRNA | AK028381.1 | - | 4 | 585 | ||
AK129051.1 | - | 4 | 593 | |||
BC030382.1 | - | 4 | 550 | |||
BC115716.1 | - | 4 | 562 | 40085888 | ||
CO433698.1 | - | 2 | 179 |