Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177372.3 | NP_796346.2 | 10 | 1625 | ||
GenBank mRNA | AK028381.1 | - | 10 | 1602 | ||
AK129051.1 | - | 10 | 1610 | |||
BC025182.1 | - | 5 | 776 | |||
BC030382.1 | - | 10 | 1566 | |||
BC115716.1 | - | 10 | 1579 | 40085888 |