Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cndp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177450.4 | NP_803233.2 | 5, 6 | 759 | ||
GenBank mRNA | AK085308.1 | - | 5, 6 | 749 | ||
AK165500.1 | - | 4, 5 | 2215 | |||
AK165706.1 | - | 5, 6 | 711 | |||
BC043305.1 | - | 5, 6 | 688 | 5040944 |