Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cndp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177450.4 | NP_803233.2 | 6 | 825 | ||
GenBank mRNA | AK085308.1 | - | 6 | 815 | ||
AK165500.1 | - | 5 | 2281 | |||
AK165706.1 | - | 6 | 777 | |||
BC043305.1 | - | 6 | 754 | 5040944 |