Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skint10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177668.2 | NP_808336.1 | 3 | 203 | ||
GenBank mRNA | AK037152.1 | - | 3 | 168 | ||
EF494905.1 | - | 3 | 166 | |||
EF494906.1 | - | 3 | 166 |