Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mcf2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159485.1 | NP_001152957.1 | 29 | 4262 | ||
NM_001159486.1 | NP_001152958.1 | 30 | 4176 | |||
NM_178076.3 | NP_835177.2 | 31 | 4333 | |||
GenBank mRNA | AK122259.1 | - | 21 | 3145 | ||
AK139245.1 | - | 1 | 798 | |||
AK147252.1 | - | 29 | 4400 | |||
AK170558.1 | - | 29 | 4196 | |||
BC029315.1 | - | 10 | 1848 | |||
BC058622.1 | - | 31 | 4268 | 5704822 |