Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldlrad3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290784.1 | NP_001277713.1 | 6 | 2205 | ||
NM_178886.3 | NP_849217.2 | 6 | 2208 | |||
GenBank mRNA | AK029683.1 | - | 6 | 2107 | ||
AK032208.1 | - | 6 | 2094 |