Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Poln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289803.1 | NP_001276732.1 | 4 | 404 | ||
NM_001289804.1 | NP_001276733.1 | 5 | 453 | |||
NM_181857.4 | NP_862905.1 | 4 | 404 | |||
GenBank mRNA | AK133063.1 | - | 5 | 373 | ||
AY135562.3 | - | 4 | 326 | |||
BC141101.1 | - | 4 | 304 | 9055713 | ||
BC145378.1 | - | 4 | 304 | 9053936 |