Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK011109.1 | - | 2 | 374 | ||
AK011167.1 | - | 2 | 476 | |||
AK020031.1 | - | 2 | 531 | |||
AK029454.1 | - | 2 | 364 | |||
AK146167.1 | - | 2 | 253 | |||
AK156717.1 | - | 3 | 796 | |||
AK168313.1 | - | 725 | ||||
BC027368.1 | - | 2 | 680 | |||
BE655013.1 | - | 2 | 282 |