Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100A12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005621.1 | NP_005612.1 | 3 | 280 | ||
GenBank mRNA | BC070294.1 | - | 3 | 280 | 30415232 | |
D49549.1 | - | 2 | 234 | |||
D83664.1 | - | 3 | 280 | |||
X97859.1 | - | 3 | 263 |