Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STK38L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015000.3 | NP_055815.1 | 3 | 376 | ||
XM_006719058.3 | XP_006719121.1 | 4 | 357 | |||
XM_006719059.3 | XP_006719122.1 | 4 | 336 | |||
XM_017019036.1 | XP_016874525.1 | 3 | 332 | |||
GenBank mRNA | AB023182.2 | - | 3 | 461 | ||
AK291055.1 | - | 3 | 266 | |||
BC028603.1 | - | 3 | 350 | 4823429 |