Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdtc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199306.4 | NP_955010.1 | 11 | 1384 | ||
GenBank mRNA | AK131700.1 | - | 13 | 1540 | ||
AK172431.1 | - | 11 | 1430 | |||
AK188298.1 | - | 4 | 238 | |||
BC034833.1 | - | 4 | 307 | 5099225 | ||
BC048824.1 | - | 11 | 1178 | 4188979 | ||
BC057107.1 | - | 10 | 1027 | 5697877 |