Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDCBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007067.1 | NP_001007068.1 | 9 | 1046 | ||
NM_001007068.1 | NP_001007069.1 | 9 | 1031 | |||
NM_001007069.1 | NP_001007070.1 | 9 | 1046 | |||
NM_001007070.1 | NP_001007071.1 | 9 | 1043 | |||
NM_005625.3 | NP_005616.2 | 9 | 1049 | |||
XM_011517571.2 | XP_011515873.2 | 9 | 2418 | |||
XM_017013727.1 | XP_016869216.1 | 9 | 2421 | |||
XM_017013728.1 | XP_016869217.1 | 9 | 2418 | |||
GenBank mRNA | AF000652.1 | - | 9 | 1047 | ||
AF006636.1 | - | 9 | 974 | |||
AK128645.1 | - | 10 | 2473 | |||
AK295198.1 | - | 8 | 786 | |||
AK298697.1 | - | 9 | 861 | |||
AK300647.1 | - | 9 | 963 | |||
BC013254.2 | - | 9 | 933 | |||
BC113674.1 | - | 9 | 993 | 8322726 | ||
BC113676.1 | - | 9 | 993 | 8322728 | ||
BC143915.1 | - | 9 | 990 | 9052433 | ||
BC143916.1 | - | 9 | 993 | 9052434 | ||
U83463.1 | - | 8 | 923 |