Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193266.1 | NP_001180195.2 | 7 | 2788 | ||
NM_207010.2 | NP_996893.3 | 7 | 2788 | |||
GenBank mRNA | AK137667.1 | - | 7 | 2789 | ||
AY371925.1 | - | 7 | 1442 | |||
BC137901.1 | - | 7 | 1441 | 8860920 | ||
BC137902.1 | - | 7 | 1441 | 8860921 | ||
BC145021.1 | - | 7 | 1436 | |||
BC171938.1 | - | 7 | 1441 | 9054224 |