Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpcat4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207206.2 | NP_997089.1 | 1 | 180 | ||
GenBank mRNA | BC068131.1 | - | 1 | 180 | 6414152 | |
BC080829.1 | - | 1 | 147 | 30435722 |