Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exoc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207214.3 | NP_997097.1 | 2 | 623 | ||
GenBank mRNA | AK134562.1 | - | 2 | 135 | ||
AK152307.1 | - | 2 | 352 | |||
AK157473.1 | - | 2 | 318 | |||
AK164067.1 | - | 2 | 345 | |||
AK167145.1 | - | 2 | 311 | |||
AK170148.1 | - | 2 | 316 | |||
AK171012.1 | - | 2 | 341 | |||
BC049967.1 | - | 2 | 280 | 3672313 |