Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptpdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301781.1 | NP_001288710.1 | 8 | 1139 | ||
NM_207232.2 | NP_997115.1 | 7 | 956 | |||
GenBank mRNA | AK030415.1 | - | 7 | 949 | ||
AK038922.1 | - | 7 | 869 | |||
AK053480.1 | - | 7 | 727 | |||
AK132781.1 | - | 8 | 1048 | |||
BC066081.1 | - | 7 | 857 | 30355705 |