Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Man1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207237.3 | NP_997120.1 | 5 | 1723 | ||
GenBank mRNA | AK141122.1 | - | 5 | 1201 | ||
AK179360.1 | - | 2 | 168 | |||
BC067023.1 | - | 6 | 1293 | 6848230 |