Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EVI5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308248.1 | NP_001295177.1 | 3, 4 | 474 | ||
NM_005665.5 | NP_005656.4 | 3, 4 | 474 | |||
XM_017002271.1 | XP_016857760.1 | 3, 4 | 472 | |||
XM_017002276.1 | XP_016857765.1 | 3, 4 | 696 | |||
XM_017002277.1 | XP_016857766.1 | 3, 4 | 472 | |||
XM_017002280.1 | XP_016857769.1 | 3, 4 | 696 | |||
XM_017002281.1 | XP_016857770.1 | 3, 4 | 774 | |||
XM_017002283.1 | XP_016857772.1 | 3, 4 | 472 | |||
XM_017002284.1 | XP_016857773.1 | 3, 4 | 695 | |||
XM_017002285.1 | XP_016857774.1 | 3, 4 | 695 | |||
XM_017002286.1 | XP_016857775.1 | 2, 3 | 466 | |||
GenBank mRNA | AB449875.1 | - | 3, 4 | 464 | ||
AF008915.1 | - | 3, 4 | 475 | |||
BP282704.1 | - | 3, 4 | 474 |