Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281812.1 | NP_001268741.1 | 3 | 164 | ||
NM_207279.2 | NP_997162.2 | 4 | 331 | |||
GenBank mRNA | AK039459.1 | - | 2 | 71 | ||
AK149435.1 | - | 3 | 165 | |||
BC034116.1 | - | 1 | 196 | |||
BC039627.1 | - | 4 | 331 | 5038680 |