Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcxd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281812.1 | NP_001268741.1 | 7 | 1171 | ||
NM_207279.2 | NP_997162.2 | 8 | 1336 | |||
GenBank mRNA | AK149435.1 | - | 7 | 1172 | ||
BC034116.1 | - | 5 | 1029 | |||
BC039627.1 | - | 8 | 1336 | 5038680 |