Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zranb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207302.1 | NP_997185.1 | 1 | 454 | ||
GenBank mRNA | AK140426.1 | - | 2 | 828 | ||
AK163589.1 | - | 2 | 895 | |||
BC145520.1 | - | 1 | 492 |