Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMK2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015981.3 | NP_057065.2 | 10 | 958 | ||
NM_171825.2 | NP_741960.1 | 10 | 958 | |||
XM_017009898.1 | XP_016865387.1 | 10 | 958 | |||
GenBank mRNA | AB023185.1 | - | 10 | 891 | ||
AF091486.1 | - | 10 | 847 | |||
AF145710.1 | - | 10 | 749 | |||
AF145711.1 | - | 10 | 749 | |||
AK289776.1 | - | 10 | 958 | |||
AK308853.1 | - | 9 | 831 | |||
AY429524.1 | - | 4 | 250 | |||
BC040457.1 | - | 10 | 873 | 4811948 |