Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005692.4 | NP_005683.2 | 2 | 376 | ||
NM_007189.2 | NP_009120.1 | 2 | 376 | |||
XM_017011639.1 | XP_016867128.1 | 2 | 253 | |||
XM_017011640.1 | XP_016867129.1 | 2 | 338 | |||
GenBank mRNA | AL050291.1 | - | 1 | 168 | ||
BC001661.2 | - | 2 | 208 | 2988125 | ||
BT007451.1 | - | 1 | 122 | |||
CR987440.1 | - | 1 | 107 | |||
HY165500.1 | - | 2 | 380 |