Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gripap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290455.1 | NP_001277384.1 | 2 | 120 | ||
NM_207670.2 | NP_997553.1 | 2 | 120 | |||
GenBank mRNA | AK156165.1 | - | 2 | 121 | ||
AK158098.1 | - | 2 | 101 | |||
AK172590.1 | - | 2 | 121 | |||
AK172623.1 | - | 2 | 121 | |||
AK172699.1 | - | 2 | 121 | |||
AW342118.1 | - | 2 | 95 | |||
BB848425.1 | - | 2 | 348 | |||
BC138523.1 | - | 2 | 116 | 8861543 | ||
BC145310.1 | - | 2 | 116 | 9053864 | ||
X97571.1 | - | 2 | 89 |