Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gripap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290455.1 | NP_001277384.1 | 26 | 2643 | ||
NM_207670.2 | NP_997553.1 | 25 | 2550 | |||
GenBank mRNA | AK156165.1 | - | 25 | 2551 | ||
AK172623.1 | - | 25 | 2551 | |||
AK172699.1 | - | 25 | 2551 | |||
AK173119.1 | - | 11 | 3024 | |||
BC035552.1 | - | 2 | 576 |