Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256670.1 | NP_001243599.1 | 3, 4 | 871 | ||
NM_005700.4 | NP_005691.2 | 4, 5 | 961 | |||
NM_130443.3 | NP_569710.2 | 4, 5 | 964 | |||
GenBank mRNA | AJ271216.1 | - | 4, 5 | 560 | ||
AK021449.1 | - | 4, 5 | 539 | |||
AK222566.1 | - | 4, 5 | 538 | |||
AK222847.1 | - | 4, 5 | 539 | |||
AK297199.1 | - | 3, 4 | 446 | |||
AK304581.1 | - | 5, 6 | 462 | |||
AK315478.1 | - | 4, 5 | 952 | |||
AL833475.1 | - | 4, 5 | 566 | |||
BC001446.2 | - | 3, 4 | 524 | 3138626 | ||
BC007221.1 | - | 4, 5 | 550 | 3349135 | ||
BC014038.2 | - | 4, 5 | 524 | 4547232 | ||
BC024271.1 | - | 3, 4 | 520 | 4564651 |