Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011153.3 | NP_035283.1 | 5 | 1188 | ||
GenBank mRNA | AF071562.1 | - | 5 | 1189 | ||
AF500906.1 | - | 5 | 1198 | |||
AK078335.1 | - | 5 | 1205 | |||
BC026822.1 | - | 5 | 1188 | 4511839 |