Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eftud2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109995.1 | NP_001103465.1 | 5, 6 | 521 | ||
NM_011431.3 | NP_035561.1 | 5, 6 | 518 | |||
GenBank mRNA | AK052242.1 | - | 5, 6 | 476 | ||
AK150158.1 | - | 5, 6 | 490 | |||
AK153473.1 | - | 5, 6 | 490 | |||
AK154878.1 | - | 5, 6 | 500 | |||
AK165151.1 | - | 5, 6 | 508 | |||
AK165624.1 | - | 5, 6 | 492 | |||
AK172875.1 | - | 5, 6 | 472 | |||
AK211645.1 | - | 1, 2 | 44 | |||
BC052674.1 | - | 5, 6 | 494 | 30061580 | ||
BC054778.1 | - | 5, 6 | 484 | 6810127 | ||
BQ886207.1 | - | 5, 6 | 513 | |||
U97079.1 | - | 5, 6 | 460 |