Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011879.2 | NP_036009.1 | 6 | 569 | ||
GenBank mRNA | AJ006130.1 | - | 6 | 452 | ||
AK029644.1 | - | 6 | 523 | |||
AK050625.1 | - | 6 | 527 | |||
AK082356.1 | - | 6 | 501 | |||
AK087976.1 | - | 6 | 515 | |||
AK088908.1 | - | 6 | 504 | |||
AK163766.1 | - | 6 | 523 | |||
AK167238.1 | - | 6 | 534 | |||
BC006706.1 | - | 5 | 699 | |||
BC014739.1 | - | 6 | 500 | 4920184 | ||
BC026569.1 | - | 6 | 500 | 4982612 | ||
BC029148.1 | - | 6 | 542 | |||
BC029671.1 | - | 6 | 512 | 5364953 | ||
BC029694.1 | - | 6 | 509 | 5369477 | ||
BC033356.1 | - | 6 | 496 | 5366230 | ||
BG084120.2 | - | 6 | 569 |