Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011986.4 | NP_036116.3 | 7 | 3286 | ||
GenBank mRNA | AK122327.1 | - | 5 | 2631 | ||
AK154889.1 | - | 7 | 3267 | |||
AK170502.1 | - | 8 | 3001 | |||
AK185185.1 | - | 2 | 175 | |||
BC017126.1 | - | 7 | 3217 | 4485839 |