Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ecsit | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253897.1 | NP_001240826.1 | 4 | 495 | ||
NM_001253898.1 | NP_001240827.1 | 3 | 317 | |||
NM_012029.2 | NP_036159.1 | 4 | 503 | |||
GenBank mRNA | AF182510.1 | - | 3 | 516 | ||
AK007564.1 | - | 4 | 477 | |||
AK012835.1 | - | 4 | 475 | |||
AK219052.1 | - | 1 | 41 | |||
BY123162.1 | - | 3 | 297 |