Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clnk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013748.3 | NP_038776.3 | 17, 18 | 1126 | ||
GenBank mRNA | AB021220.1 | - | 17, 18 | 1246 | ||
AF187819.1 | - | 17, 18 | 1126 | |||
AK082826.1 | - | 13, 14 | 864 | |||
BC125637.1 | - | 17, 18 | 1123 | 40130152 | ||
BC125639.1 | - | 17, 18 | 1123 | 40130154 |