Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elane | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015779.2 | NP_056594.2 | 3 | 376 | ||
GenBank mRNA | AK143710.1 | - | 3 | 376 | ||
BC145800.1 | - | 3 | 441 | 40131103 |