Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clcf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019952.3 | NP_064336.1 | 3 | 437 | ||
GenBank mRNA | AF176913.1 | - | 3 | 438 | ||
AK137396.1 | - | 2298 | ||||
AK166218.1 | - | 626 | ||||
AK166421.1 | - | 442 | ||||
AK170969.1 | - | 3 | 455 | |||
BC104258.1 | - | 3 | 412 | 40039794 | ||
BC104259.1 | - | 3 | 412 | 40039795 |