Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jmy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021310.3 | NP_067285.2 | 1 | 1404 | ||
GenBank mRNA | AB221690.1 | - | 1 | 902 | ||
AF201390.1 | - | 1 | 1028 | |||
AK139934.1 | - | 1 | 1033 | |||
AK141957.1 | - | 1 | 1405 | |||
BC020052.1 | - | 1 | 1329 | |||
BC069906.1 | - | 1 | 1258 | |||
BC090835.1 | - | 1 | 1285 | 5705427 |