Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287164.1 | NP_001274093.1 | 3, 4 | 366 | ||
NM_001287165.1 | NP_001274094.1 | 3, 4 | 366 | |||
NM_001287166.1 | NP_001274095.1 | 3, 4 | 366 | |||
NM_001287167.1 | NP_001274096.1 | 3, 4 | 346 | |||
NM_022979.2 | NP_075355.1 | 3, 4 | 366 | |||
GenBank mRNA | AF201386.1 | - | 2, 3 | 152 | ||
AK089973.1 | - | 3, 4 | 353 | |||
AK137234.1 | - | 3, 4 | 347 | |||
AK146491.1 | - | 3, 4 | 347 | |||
AK164402.1 | - | 3, 4 | 351 | |||
AK164743.1 | - | 1, 2 | 67 | |||
AK172207.1 | - | 3, 4 | 343 | |||
BC050911.1 | - | 3, 4 | 320 | 6417631 | ||
BC057608.1 | - | 3, 4 | 320 | 6417631 | ||
BC112912.1 | - | 3, 4 | 320 | 5697919 | ||
BC137975.1 | - | 2, 3 | 225 | 8860994 | ||
BC145071.1 | - | 2, 3 | 225 | 9053619 |