Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARPC4-TTLL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198793.1 | NP_001185722.1 | 2, 3 | 704 | ||
GenBank mRNA | AK024110.1 | - | 2, 3 | 694 | ||
BC025289.1 | - | 1, 2 | 138 |
Gene Symbol: ARPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024959.2 | NP_001020130.1 | 2, 3 | 338 | ||
NM_001024960.2 | NP_001020131.1 | 2, 3 | 193 | |||
NM_001198780.1 | NP_001185709.1 | 2, 3 | 192 | |||
NM_005718.4 | NP_005709.1 | 2, 3 | 704 | |||
GenBank mRNA | AF006087.1 | - | 2, 3 | 145 | ||
AF019888.1 | - | 1, 2 | 130 | |||
AW408422.1 | - | 2, 3 | 151 | |||
BC012596.2 | - | 1, 2 | 127 | |||
BG938423.1 | - | 2, 3 | 176 | |||
BI768875.1 | - | 2, 3 | 317 | |||
BI910730.1 | - | 2, 3 | 712 | |||
BM045872.1 | - | 2, 3 | 203 | |||
BQ688329.1 | - | 2, 3 | 276 | |||
BX419672.2 | - | 2, 3 | 192 | |||
DA226584.1 | - | 2, 3 | 193 | |||
DB060690.1 | - | 2, 3 | 338 |