Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159941.1 | NP_001153413.1 | 5, 6 | 615 | ||
NM_026145.4 | NP_080421.2 | 5, 6 | 612 | |||
GenBank mRNA | AK028657.1 | - | 5, 6 | 579 | ||
AK041516.1 | - | 5, 6 | 579 | |||
AK145295.1 | - | 5, 6 | 591 | |||
AK147714.1 | - | 5, 6 | 557 | |||
AK161199.1 | - | 5, 6 | 569 | |||
AK163351.1 | - | 5, 6 | 566 | |||
AK166505.1 | - | 5, 6 | 591 | |||
AK216751.1 | - | 2, 3 | 105 | |||
BC006935.1 | - | 5, 6 | 593 | 3601917 | ||
BQ571808.1 | - | 5, 6 | 566 |