Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029545.2 | NP_083821.1 | 3 | 2095 | ||
GenBank mRNA | AK032228.1 | - | 3 | 2092 | ||
AK039019.1 | - | 1114 | ||||
AK042771.1 | - | 3 | 2096 |