Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crispld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031402.2 | NP_113579.2 | 12 | 1624 | ||
GenBank mRNA | AF329198.1 | - | 12 | 1621 | ||
AK132405.1 | - | 12 | 1362 | |||
AK133870.1 | - | 12 | 1625 | |||
AK134273.1 | - | 12 | 1560 | |||
AK137611.1 | - | 12 | 1625 | |||
BC034334.1 | - | 3 | 229 | |||
BC040768.1 | - | 12 | 1953 | 4458200 |