Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam126a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053090.2 | NP_444320.2 | 8 | 921 | ||
GenBank mRNA | AK036680.1 | - | 1 | 1825 | ||
AK155209.1 | - | 8 | 922 | |||
AK161265.1 | - | 8 | 1115 | |||
BC056381.1 | - | 8 | 860 | 6852011 | ||
BC060692.1 | - | 8 | 908 | 6410470 |