Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam126a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053090.2 | NP_444320.2 | 11 | 3570 | ||
GenBank mRNA | AK136511.1 | - | 1 | 425 | ||
BC056381.1 | - | 11 | 3508 | 6852011 | ||
BC060692.1 | - | 11 | 3557 | 6410470 |