Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053109.3 | NP_444339.1 | 5 | 616 | ||
GenBank mRNA | AF321553.1 | - | 5 | 609 | ||
AF350409.1 | - | 5 | 601 | |||
AY320031.1 | - | 5 | 616 | |||
BC106776.1 | - | 5 | 576 | 40040181 | ||
HQ713439.1 | - | 5 | 550 | |||
HQ875062.1 | - | 5 | 550 |