Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053259.2 | NP_444489.2 | 5 | 688 | ||
GenBank mRNA | AF184895.1 | - | 4 | 527 | ||
AK144498.1 | - | 5 | 689 | |||
BC119386.1 | - | 4 | 555 | 8734135 | ||
BC119388.1 | - | 4 | 555 | 8734137 |