Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd209c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130903.3 | NP_570973.1 | 2 | 1067 | ||
GenBank mRNA | AF373410.1 | - | 2 | 1067 | ||
BC120804.1 | - | 2 | 257 | 40129727 | ||
BC132635.1 | - | 2 | 257 | 40130912 | ||
BC145088.1 | - | 2 | 257 | 9053636 | ||
BC145089.1 | - | 2 | 257 | |||
BC145090.1 | - | 1 | 365 |