Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005727.3 | NP_005718.2 | 7 | 959 | ||
XM_011540460.2 | XP_011538762.1 | 7 | 933 | |||
GenBank mRNA | AF054838.1 | - | 5 | 485 | ||
AF065388.1 | - | 6 | 606 | |||
AF133425.1 | - | 6 | 608 | |||
AK313774.1 | - | 7 | 949 | |||
BC007290.1 | - | 6 | 612 | 3349295 | ||
BC013404.2 | - | 6 | 610 | 2959379 |