Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txlnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138628.3 | NP_619534.2 | 3 | 666 | ||
GenBank mRNA | AF422244.1 | - | 3 | 603 | ||
AF422245.1 | - | 3 | 603 | |||
AK052324.1 | - | 3 | 630 | |||
AK084579.1 | - | 3 | 666 | |||
AK084639.1 | - | 3 | 667 | |||
AK085904.1 | - | 3 | 560 | |||
AK137442.1 | - | 3 | 581 | |||
BC141023.1 | - | 3 | 579 | 9055635 |