Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPHB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001309192.1 | NP_001296121.1 | 14 | 3132 | ||
NM_001309193.1 | NP_001296122.1 | 17 | 3213 | |||
NM_004442.7 | NP_004433.2 | 16 | 3309 | |||
NM_017449.4 | NP_059145.2 | 16 | 3306 | |||
XM_006710441.3 | XP_006710504.1 | 16 | 3402 | |||
XM_006710442.3 | XP_006710505.1 | 16 | 3284 | |||
GenBank mRNA | AB210018.1 | - | 16 | 3215 | ||
AF025304.1 | - | 17 | 3093 | |||
BC018763.1 | - | 14 | 2459 | |||
L41939.1 | - | 16 | 3268 |